Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper (II) Sulfate, 1 M Solution, Innovating Science™
SCI_ED

Catalog No. S031072 Shop All Aldon Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S031072 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S031072 Supplier Aldon Corporation Supplier No. IS12196
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use 1.0 Molar solution for laboratory and industrial chemical applications.

Chemical Identifiers

CAS 7732-18-5 , 7758-99-8

Specifications

Physical Form Liquid or Solid
Quantity 500 mL
Grade Reagent
Concentration or Composition (by Analyte or Components) 1 M
Packaging HDPE
Chemical Name or Material Cupric Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.