Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Copper(II) sulfate | 7758-98-7 | 100.4% | 159.60 | 100 G
SDP

Catalog No. 5000364953
Encompass_Preferred

Copper(II) sulfate, also known as cupric sulfate, is an inorganic compound that provides copper ions. It is utilized in forming coordination compounds with ligands and can be applied in various enzymatic studies and material synthesis. This compound is also suitable for synthesizing anionic copper sulfate aggregates and chains for magnetic research, and as a copper supplement for evaluating bioavailability in animal nutrition.

  • Source of copper ions
  • Forms coordination compounds with ligands
  • Used for enzymatic studies and material synthesis
  • Synthesizes anionic copper sulfate aggregates and chains for magnetic research
  • Functions as a copper supplement in animal nutrition
  • Molecular formula: CuSO4
  • Appearance: White to off-white solid
  • Storage: Store at room temperature, keep dry and cool

Catalog No. 50-003-64953 Supplier Medchemexpress LLC Supplier No. 100GHYY1878
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Copper(II) sulfate, also known as cupric sulfate, is an inorganic compound that provides copper ions. It is utilized in forming coordination compounds with ligands and can be applied in various enzymatic studies and material synthesis. This compound is also suitable for synthesizing anionic copper sulfate aggregates and chains for magnetic research, and as a copper supplement for evaluating bioavailability in animal nutrition.

  • Source of copper ions
  • Forms coordination compounds with ligands
  • Used for enzymatic studies and material synthesis
  • Synthesizes anionic copper sulfate aggregates and chains for magnetic research
  • Functions as a copper supplement in animal nutrition
  • Molecular formula: CuSO4
  • Appearance: White to off-white solid
  • Storage: Store at room temperature, keep dry and cool

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.