Learn More
Medchemexpress LLC Copper(II) sulfate | 7758-98-7 | 100.4% | 159.60 | 25 G
SDP
Catalog No. 5000365671
Encompass_Preferred
Copper(II) sulfate is an inorganic compound that serves as a source of copper ions. It can form coordination compounds when copper ions coordinate with ligands. This compound is utilized for various enzymatic studies and in material synthesis.
  • Serves as a source of copper ions
  • Forms coordination compounds
  • Utilized for various enzymatic studies
  • Used in material synthesis
  • Synthesizes anionic copper sulfate aggregates and chains for magnetic research
  • Employed as a copper supplement in animal nutrition
Catalog No. 50-003-65671 Supplier Medchemexpress LLC Supplier No. 25GHYY1878
Copper(II) sulfate is an inorganic compound that serves as a source of copper ions. It can form coordination compounds when copper ions coordinate with ligands. This compound is utilized for various enzymatic studies and in material synthesis.
  • Serves as a source of copper ions
  • Forms coordination compounds
  • Utilized for various enzymatic studies
  • Used in material synthesis
  • Synthesizes anionic copper sulfate aggregates and chains for magnetic research
  • Employed as a copper supplement in animal nutrition