Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(Ii) Sulfate, Honeywell Fluka™

Catalog No. 6001956 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
6 x 250 g
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-56 1 kg
60-019-57 250 g
60-044-767 6 x 250 g
60-044-766 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-019-56 Supplier Honeywell Chemicals Supplier No. 128521KG
Only null left
Add to Cart
Add to Cart

Puriss., anhydrous, meets analytical specification of BP, Ph. Eur., USP, 99-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula CuSO4
UN Number 3077
Formula Weight 159.61g/mol
Percent Purity 99 to 100.5% (based on anhydrous substance)
Grade puriss.
Vapor Pressure 7.3mmHg (25°C)
Density 3.603g/mL at 25°C
Chemical Name or Material Copper(Ii) Sulfate, Meets analytical specification of Ph. Eur.BP, USP
Hazard Statement P301 + P312 + P330-P305 + P351 + P338
Precautionary Statement H302-H315-H319-H410
EINECSNumber 231-847-6
RTECSNumber GL8800000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.