Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA1070122
Click to view available options
Quantity:
25 g
100 g

CAS: 23254-43-5 | CuO4S | 159.60 (anhydrous basis) g/mol

It is used as a catalyst in certain organic transformations like synthesis of ynamides. It is used in Fehling's solution and Benedict's solution for testing reducing sugars, in the Biuret reagent for testing proteins, and in combination with potassium permanganate for testing blood for anemia. It is used in the preparation of Cheshunt compound for horticulture. It finds use as a coloring ingredient in artworks, as an oxidant in combination with potassium permanganate for oxidizing primary alcohols, and as a mordant in dyeing. It is used to etch copper or zinc plates for intaglio print making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23254-43-5
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.60 (anhydrous basis)
MDL Number MFCD00149682
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate monohydrate, cupric sulfate monohydrate, copper sulfate, monohydrate, copper sulfate hydrate, caswell no. 257, epa pesticide chemical code 024402, copper ii sulfate hydrate, sulfuric acid copper 2+ salt 1:1 , monohydrate, copper sulphate hydrate, cuso4.h2o
PubChem CID 61499
IUPAC Name copper;sulfate;hydrate
SMILES [Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
Physical Form Crystalline Aggregates
Quantity 100 g
Assay Percent Range 99.999% (Metal basis)
Solubility Information Soluble in water and methanol. Insoluble in ethanol and most of the organic solvents.
Formula Weight 159.60 (Anhydrous)
Odor Odorless
Appearance Blue crystalline aggregrates
Chemical Name or Material Copper(II) sulfate hydrate, m.p. 30° -2H2O, 110° to4H2O, 250° anhydrous; 2.286
Hazard Category H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation DOT Class: 9, Packaging Group: III
EINECSNumber 231-847-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.