Learn More
Strem, An Ascensus Company CAS# 7758-99-8. 25g. Copper(II) sulfate pentahydrate (99.999%-Cu) PURATREM. MFCD00149681
SDP
Catalog No. 5090114910
Encompass
CAS# 7758-99-8. 25g. Copper(II) sulfate pentahydrate (99.999%-Cu) PURATREM. MFCD00149681. Molecular Weight: 159.60 (249.68). Molecular Formula: CuSO4∙5H2O. Color/form: blue xtl. Strem# 93-2959. http://www.strem.com/catalog/v/93-2959/
Catalog No. 50-901-14910 Supplier Strem, An Ascensus Company Supplier No. 93295925G
CAS# 7758-99-8. 25g. Copper(II) sulfate pentahydrate (99.999%-Cu) PURATREM. MFCD00149681. Molecular Weight: 159.60 (249.68). Molecular Formula: CuSO4∙5H2O. Color/form: blue xtl. Strem# 93-2959. http://www.strem.com/catalog/v/93-2959/