Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) sulfate pentahydrate

Catalog No. 46913050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-913-050G 50 g
46-913-0250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 46-913-050G Supplier Sigma Aldrich Supplier No. 46913050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(II) sulfate pentahydrate

Specifications

CAS 7758-99-8
Melting Point 110°C
Molecular Formula CuO4S -+ 5H2O
Linear Formula CuSO4 -+ 5H2O
Quantity 50 g
MDL Number MFCD00149681
Synonym Blue Vitriol; Cupric sulfate pentahydrate; Sulfuric acid, copper(II) salt (1:1) pentahydrate
Molecular Weight (g/mol) 249.69
Percent Purity 99.995% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.