Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate pentahydrate, ≥98.0%, Reagent Grade, Solstice

Catalog No. 60047915 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-915 500 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-915 Supplier Solstice Supplier No. C7631500G

missing translation for 'validMainframeMsg'

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper;sulfate;pentahydrate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Melting Point 110°C
Color Blue Green
pH 3.5 to 4.5 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula CuSO4 · 5H2O
UN Number UN3077
Formula Weight 249.69g/mol
Percent Purity ≥98.0%
Odor Odorless
Packaging Poly bottle
Vapor Pressure 7.3mmHg (25°C)
Density 2.28g/cu.cm at 20°C
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less
Hazard Statement P273-P301 + P312 + P330-P305 + P351 + P338-P391-P501
DOTInformation 9/PGIII
RTECSNumber 231-847-6
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.