Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate pentahydrate cryst. EMPROVE™ ESSENTIAL Ph Eur, BP, USP, 100.5%, MilliporeSigma™

Catalog No. M1027871000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1027871000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1027871000 Supplier MilliporeSigma Supplier No. 1027871000

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7758-99-8
Molecular Formula CuO4S * 5 H2O
Molecular Weight (g/mol) 249.68
Synonym Copper monosulfate pentahydrate, Copper vitriol pentahydrate

Specifications

Melting Point 147°C
pH 3.5 to 4.5 (50 g/L, H2O, 20°C)
Quantity 1 kg
Assay Percent Range Assay (iodometric): 99.0 to 100.5%; Assay (iodometric, dried substance): 98.5 to 100.5%
Solubility Information 317 g/L
Packaging Plastic Bottle
Density 2.284 g/cm3 (at 20°C)
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.