Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper(II) sulfate solution, Volumetric, 0.1 M CuSO4, Solstice

Catalog No. 60046540 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-540 6 x 1 L
60-004-51 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-540 Supplier Solstice Supplier No. 351856X1L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.1 M CuSO4

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7758-98-7 , 7732-18-5
Molecular Formula CuO4S
Molecular Weight (g/mol) 159.602
MDL Number MFCD00010981
InChI Key ARUVKPQLZAKDPS-UHFFFAOYSA-L
Synonym copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
PubChem CID 24462
ChEBI CHEBI:23414
IUPAC Name copper;sulfate
SMILES [O-]S(=O)(=O)[O-].[Cu+2]

Specifications

Boiling Point >100°C
CAS Min % ≥90.0000%, ≥0.2500%
Physical Form Liquid
CAS Max % ≤100.0000%, <2.5000%
Quantity 6 x 1 L
Packaging Poly Bottle
Density 1.02 g/cm3
UN Number UN3082
Formula Weight 159.61
Concentration or Composition (by Analyte or Components) 0.1 M CuSO4
Chemical Name or Material Copper(II) sulfate solution, Volumetric, 0.1 M CuSO4
Show More Show Less
Hazard Statement P273-P501
ShelfLife 450 days from date of manufacture
DOTInformation 9 / PGIII
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.