Learn More
Lumiprobe Copper(II)-TВTA catalytic buffer, 1.5x, 55 mL
Catalog No. NC3360275
Encompass_Preferred
Ready-to-use catalytic buffer for click chemistry (pH 7.0) containing copper(II), TBTA, triethylammonium acetate, and DMSO. The buffer has a concentration of 1.5×. This reagent is suitable for modifying nucleic acids such as oligonucleotides and small molecules.Copper(II) should be reduced to copper(I) immediately before reaction. Ascorbic acid is a recommended reductant.
Catalog No. NC3360275 Supplier Lumiprobe Supplier No. 61150
Ready-to-use catalytic buffer for click chemistry (pH 7.0) containing copper(II), TBTA, triethylammonium acetate, and DMSO. The buffer has a concentration of 1.5×. This reagent is suitable for modifying nucleic acids such as oligonucleotides and small molecules.Copper(II) should be reduced to copper(I) immediately before reaction. Ascorbic acid is a recommended reductant.