Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper(II) tetrafluoroborate hydrate

Catalog No. 36658750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-658-750G 50 g
36-658-7250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-658-750G Supplier Sigma Aldrich Supplier No. 36658750G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper(II) tetrafluoroborate hydrate may be used as catalyst for chemoselectiveN-tert-butoxycarbonylation of amines with di-tert-butyl dicarbonate.The product is a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

Specifications

CAS 207121-39-9
Molecular Formula B2CuF8 -+ xH2O
Linear Formula Cu(BF4)2 -+ xH2O
Quantity 50 g
MDL Number MFCD00150223
Molecular Weight (g/mol) 237.16
Percent Purity ≤25.8%Cu basis (EDTA titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.