Learn More
Electron Microscopy Sciences Omniprobe Lift-Out Grids for in-situ FIB, Copper
SDP
Catalog No. 5033768
Encompass
Custom copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both  vertical bars and "V" shaped attachment surfaces. 3 mm diameter
Material: Copper
Catalog No. 50-337-68 Supplier Electron Microscopy Sciences Supplier No. 7596401
Custom copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations with both  vertical bars and "V" shaped attachment surfaces. 3 mm diameter
Material: Copper