Learn More
Ted Pella Inc Copper/Nickel Tape, 8mm W x 20m L, each
SDP
Catalog No. NC1641336
Encompass
SEM conductive tape, electrically conductive with a clean, smooth background. Nickel is embedded in the adhesive.
Catalog No. NC1641336 Supplier Ted Pella Inc Supplier No. 16067
SEM conductive tape, electrically conductive with a clean, smooth background. Nickel is embedded in the adhesive.