Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Copper - O.F.H.C.

Catalog No. 503127096 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-7096 1 Ea.
50-312-6866 5 Ea.
50-312-6755 10 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-312-7096 Supplier Sigma Aldrich Supplier No. GF847714011EA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Copper - O.F.H.C.

Specifications

CAS 7440-50-8
Melting Point 1083.4°C
Density 8.94 g/mL (at 25°C)
Boiling Point 2567°C
Molecular Formula Cu
Linear Formula Cu
Quantity 1 Ea.
MDL Number MFCD00010965
Synonym Copper; CV000565
Molecular Weight (g/mol) 63.55
Percent Purity 99.95+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.