Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper plate, Oxygen-Free High Conductivity (OFHC), alloy 101, 2.4mm (0.093in) thick

Catalog No. p-7059704
Change view
Click to view available options
Quantity:
150 x 150 mm
300 x 300 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45210GT 150 x 150 mm
AA45210HB 300 x 300 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45210GT Supplier Thermo Scientific Chemicals Supplier No. 045210GT
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper plate, Alloy 101
Name Note Oxygen-Free High Conductivity (OFHC), 2.4mm (0.093 in.) thick
Quantity 150 x 150 mm
Form Plate
MDL Number MFCD00010965

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.