Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Reagent Powder Packs, Lovibond™

Catalog No. 09534709 Shop All Tintometer Group Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-534-709 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-709 Supplier Tintometer Group Supplier No. 530303
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-quality powder reagent for fast and easy copper measurement in water analysis.

  • These Vario powder reagents are designed for fast and efficient water analysis testing. Produced to high quality standards, they offer a reliable alternative to existing measurement systems.
  • Application: Water analysis
  • Parameter: Copper, free (HR)
  • Measurement Range: 0 to 5 mg/L Cu
  • Method Number: M2519
  • Features: Easy to use and rapid dissolution
  • Composition: Mixture of organic and inorganic compounds

Specifications

Quantity 10 mL
Type Copper reagent powder packs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.