Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach™ Copper Reagent Powder Pillow Set

Catalog No. 501996317 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Type:
Porphyrin 2 Reagent
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
50-199-6317 Porphyrin 2 Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-6317 Supplier Hach™ Supplier No. 2603649
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Set of powder reagents in individual packets for Copper determination. Hach™ Copper Reagent Powder Pillow Set includes Copper Masking Reagent, Porphyrin 1 Reagent and Porphyrin 2 Reagent. Replacement porphyrin reagents available separately for replenishment after preparing blanks.

  • Ready-to-use, premeasured powder reagents in individual sealed packets
  • For Copper determination by the Porphyrin Method (Hach Method 8143)
  • Contains sets of copper masking and porphyrin 1/2 reagent powder pillows for 100 tests
  • Porphyrin 1 and Porphyrin 2 powder pillows available separately
TRUSTED_SUSTAINABILITY

Specifications

Analytes Copper
Format Powdered Reagent
For Use With (Application) Copper determination
For Use With (Equipment) DR 6000, DR/4000, DR 3900, DR 2800, ,DR 2700, DR/2010, DR/890
Includes Porphyrin 2 Reagent Powder Pillows
No. of Tests 50 to 100 (depending on the number of blanks performed)
Quantity 1 Set
Range 2 to 210μg/L Cu
Sample Size 10 mL
Test Method Porphyrin
Type Porphyrin 2 Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.