Learn More
TA Instruments Copper Sample Pans 80µL Pkg of 100, for Use in Discovery TGA 55, 550, 5500, Ambient to 1000°C, Stainless Steel Bail Required P/N 957364.901
Catalog No. 5000580891 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Copper Sample Pans 80 µL Pkg Of 100 are for use in the Discovery TGA 55, 550, and 5500 over the temperature range of ambient to 1000°C. A stainless steel bail P/N 957364.901 is required to use the copper sample pan in a TGA.
Catalog No. 50-005-80891 Supplier TA Instruments Supplier No. 957454.901
Waters TA Instruments Copper Sample Pans 80 µL Pkg Of 100 are for use in the Discovery TGA 55, 550, and 5500 over the temperature range of ambient to 1000°C. A stainless steel bail P/N 957364.901 is required to use the copper sample pan in a TGA.