Learn More
TA Instruments Copper Sample Pans, DSC Copper Pans, Oxidative Stability Experiments, Polymers, Wire and Cable Coatings, High Thermal Conductivity, 200 Pack
Catalog No. 5000580705 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Copper Sample Pans are DSC copper pans used for oxidative stability experiments, particularly for polymeric materials used as wire and cable coatings. They offer high thermal conductivity for accurate DSC measurements.
Catalog No. 50-005-80705 Supplier TA Instruments Supplier No. 900867.901
Waters TA Instruments Copper Sample Pans are DSC copper pans used for oxidative stability experiments, particularly for polymeric materials used as wire and cable coatings. They offer high thermal conductivity for accurate DSC measurements.