Learn More
Glass Expansion Copper Skimmer Cone for Shimadzu 2030 ICP-MS (211-90200-41, 220-95356-83)
SDP
Catalog No. 501907698
Encompass_Preferred
Copper Skimmer Cone ¿ For Shimadzu 2030 ICP-MS (211-90200-41, 220-95356-83). Standard low-cost option. Not recommended for samples containing hydrofluoric acid (HF). Replace skimmer cone screws if corroded or worn (SZ5002). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number 501907703. For routine samples with better heat transfer, we recommend nickel skimmer with copper base, see catlog number 501907700.Use Nickel Skimmer for routine samples, see catalog number 501907699.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7698 Supplier Glass Expansion Supplier No. SZ7002CU
Copper Skimmer Cone ¿ For Shimadzu 2030 ICP-MS (211-90200-41, 220-95356-83). Standard low-cost option. Not recommended for samples containing hydrofluoric acid (HF). Replace skimmer cone screws if corroded or worn (SZ5002). Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For samples containing HF, and other highly corrosive samples where acid concentration begin to exceed 5%, we recommend a platinum cone, see catalog number 501907703. For routine samples with better heat transfer, we recommend nickel skimmer with copper base, see catlog number 501907700.Use Nickel Skimmer for routine samples, see catalog number 501907699.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001