Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Copper SMART 3 Reagent System
SDPSCI_ED

Catalog No. S94991 Shop All LaMotte Company Products
Catalog No. S94991 Supplier LaMotte™ Supplier No. 3640SC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Copper low range reagent system for use with SMART 3 Colorimeter.

  • Range 0.04 to 3.50 ppm
  • 50 Tests
  • Bicinchoninic Acid Method

Specifications

pH Range 6 to 8
For Use With (Application) For Industrial Waters
For Use With (Equipment) SMART 3 Colorimeter
Range 0.04 to 3.50 ppm
Test Method Bicinchoninic Acid Method
Type Reagent System
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.