Learn More
Copper Standard, Quantitative, Inorganic, 125 mL
Specifications
Specifications
| CAS | 7440-50-8, 7697-37-2, 7732-18-5 |
| CAS Max % | 97.9, 2.0, 0.1 |
| Analyte or Component Names | Water,Nitric acid,Copper |
| Molecular Formula | Cu |
| MDL Number | MFCD00010965 |
| InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
| Element | Nitric Acid, Cu |
| SMILES | [Cu] |
| Solvent or Matrix | Water/Dilute Nitric Acid |
| IUPAC Name | copper |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.