Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Standard, 50 ppm Cu in dil. HNO3, Ricca Chemical
SDP

Catalog No. R22920004A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R22920004A 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R22920004A Supplier Ricca Chemical Company Supplier No. R22920004A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7758-99-8 , 7697-37-2 , 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 97.0, 1.25, 0.02
CAS Max % 100.0, 1.3, 0.02
Physical Form Liquid
Color Blue to Colorless
Melting Point 0.0°C
Boiling Point 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 4 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1mL = 50μg Cu, 50ppm Cu, in dilute Nitric Acid
Chemical Name or Material Copper Standard
Show More Show Less
ShelfLife 12 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.