Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Standard, 200 ppm Cu, Ricca Chemical
SDP

Catalog No. 229516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-951-6 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-951-6 Supplier Ricca Chemical Company Supplier No. 229516
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 7732-18-5, 7697-37-2, 7440-50-8
Packaging Natural Poly Bottle
Molecular Formula CuH10O9S
MDL Number MFCD00149681
Color Colorless to Light Blue
Quantity 500 mL
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
IUPAC Name copper(2+) pentahydrate sulfate
Molecular Weight (g/mol) Mixture
PubChem CID 24463
ChEBI CHEBI:31440
Percent Purity 200 ppm
Grade Laboratory
Chemical Name or Material Copper standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.