Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate, 1% (w/v), Ricca Chemical
SDP

Catalog No. R23168001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R23168001A 1 L Natural Poly Bottle
R2316800500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R23168001A Supplier Ricca Chemical Company Supplier No. R23168001A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7758-99-8
Molecular Formula CuH10O9S
Molecular Weight (g/mol) 249.68
MDL Number MFCD00149681
InChI Key JZCCFEFSEZPSOG-UHFFFAOYSA-L
Synonym copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
PubChem CID 24463
ChEBI CHEBI:31440
IUPAC Name copper(2+) pentahydrate sulfate
SMILES O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

Specifications

Color Blue
CAS Min % 97.0, 0.97
CAS Max % 100.0, 1.01
Physical Form Liquid
Quantity 1 L
Percent Purity 1% W/V
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Chemical Name or Material Copper Sulfate
ShelfLife 24 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.