Learn More
Medchemexpress LLC Copper(Ⅱ) sulfate | 7758-98-7 | 100.4% | 159.60 | 50 G
SDP
Catalog No. 5000365803
Encompass_Preferred
Copper(Ⅱ) sulfate is an inorganic compound that serves as a source of copper ions, forming coordination compounds with various ligands. It is utilized in various enzymatic studies and for material synthesis.
  • Can be used to synthesize anionic copper sulfate aggregates and chains for magnetic research.
  • Can be used in enzymatic reactions and material synthesis.
  • Can be used as a copper supplement in animal nutrition to evaluate bioavailability.
Catalog No. 50-003-65803 Supplier Medchemexpress LLC Supplier No. 50GHYY1878
Copper(Ⅱ) sulfate is an inorganic compound that serves as a source of copper ions, forming coordination compounds with various ligands. It is utilized in various enzymatic studies and for material synthesis.
  • Can be used to synthesize anionic copper sulfate aggregates and chains for magnetic research.
  • Can be used in enzymatic reactions and material synthesis.
  • Can be used as a copper supplement in animal nutrition to evaluate bioavailability.