Learn More
Medchemexpress LLC Copper sulfate anhydrous | 7758-98-7 | 100.4% | 159.60 | 1 KG
SDP
Catalog No. 5000365186
Encompass_Preferred
Copper(Ⅱ) sulfate (Cupric sulfate) is an inorganic compound that serves as a source of copper ions. It forms coordination compounds by coordinating copper ions with ligands. This compound can be utilized in various enzymatic studies and for material synthesis.
  • Can be used to synthesize anionic copper sulfate aggregates and chains for magnetic research.
  • Can be used in enzymatic reactions and material synthesis.
  • Can be used as a copper supplement in animal nutrition to evaluate bioavailability.
  • For research use only.
Catalog No. 50-003-65186 Supplier Medchemexpress LLC Supplier No. 1KGHYY1878
Copper(Ⅱ) sulfate (Cupric sulfate) is an inorganic compound that serves as a source of copper ions. It forms coordination compounds by coordinating copper ions with ligands. This compound can be utilized in various enzymatic studies and for material synthesis.
  • Can be used to synthesize anionic copper sulfate aggregates and chains for magnetic research.
  • Can be used in enzymatic reactions and material synthesis.
  • Can be used as a copper supplement in animal nutrition to evaluate bioavailability.
  • For research use only.