Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Copper Sulfate, 10% (w/v), Ricca Chemical
SDP

Catalog No. 232032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
2320-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 2320-32 Supplier Ricca Chemical Company Supplier No. 232032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meets EPA Specifications

Specifications

pH Range ∼ 4 (at 0.2 Molar)
Volume (Metric) 1 L
Packaging Natural Poly Bottle
Quantity 1 L
Type COPPER SULFATE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.