Learn More
Perkin Elmer US LLC Copper Torch Ignitor Tape for Optima 8x00
Catalog No. NC0273570
Encompass_Preferred
This copper ignitor tape is also included in the Scott/Cross-Flow II Sample Introduction Kit (N0780616) and the Cyclonic/Concentric Sample Introduction Kit (N0780617) for Optima 8x00.
Catalog No. NC0273570 Supplier Perkin Elmer US LLC Supplier No. N0781097
This copper ignitor tape is also included in the Scott/Cross-Flow II Sample Introduction Kit (N0780616) and the Cyclonic/Concentric Sample Introduction Kit (N0780617) for Optima 8x00.