Learn More
TARGETMOL CHEMICALS INC Copper tripeptide 500MG
SDP
Catalog No. 502877310
Encompass_Preferred
Also available in 100 mg and bulk. Please contact Fisher for quotes. Copper tripeptide (GHK-Cu) is a naturally occurring copper complex of the tripeptide glycyl-L-histidyl-L-lysine, with a strong affinity for copper(II). First isolated from human plasma, it is also found in saliva and urine. Purity 99.94%
Catalog No. 50-287-7310 Supplier TARGETMOL CHEMICALS INC Supplier No. TP1158500MG
Also available in 100 mg and bulk. Please contact Fisher for quotes. Copper tripeptide (GHK-Cu) is a naturally occurring copper complex of the tripeptide glycyl-L-histidyl-L-lysine, with a strong affinity for copper(II). First isolated from human plasma, it is also found in saliva and urine. Purity 99.94%