Learn More
Medchemexpress LLC Copper tripeptide | 89030-95-5 | 401.91 | 250 MG
SDP
Catalog No. 5000426950
Encompass_Preferred
Copper tripeptide (GHK-Cu) is a tripeptide that may be released from existing extracellular proteins via proteolysis during wound healing. It serves as a chemoattractant for inflammatory and endothelial cells. It has been shown to increase messenger RNA production for collagen, elastin, proteoglycans, and glycosaminoglycans in fibroblasts. This peptide is a natural modulator of multiple cellular pathways in skin regeneration.
  • Acts as a chemoattractant for inflammatory and endothelial cells
  • Increases messenger RNA production for collagen, elastin, proteoglycans, and glycosaminoglycans in fibroblasts
  • Modulates multiple cellular pathways in skin regeneration
  • Affects irradiated fibroblasts, normalizing population-doubling times
  • Produces more basic fibroblast growth factors in irradiated fibroblasts
Catalog No. 50-004-26950 Supplier Medchemexpress LLC Supplier No. 250MGHYP0063
Copper tripeptide (GHK-Cu) is a tripeptide that may be released from existing extracellular proteins via proteolysis during wound healing. It serves as a chemoattractant for inflammatory and endothelial cells. It has been shown to increase messenger RNA production for collagen, elastin, proteoglycans, and glycosaminoglycans in fibroblasts. This peptide is a natural modulator of multiple cellular pathways in skin regeneration.
  • Acts as a chemoattractant for inflammatory and endothelial cells
  • Increases messenger RNA production for collagen, elastin, proteoglycans, and glycosaminoglycans in fibroblasts
  • Modulates multiple cellular pathways in skin regeneration
  • Affects irradiated fibroblasts, normalizing population-doubling times
  • Produces more basic fibroblast growth factors in irradiated fibroblasts