Learn More
Sigma Aldrich Fine Chemicals Biosciences CopperI chloride reagent g
Catalog No. 502899250
Encompass
Cu is present in 1 (cuprous) valence state in CuCl. CuCl is a semiconductor having a wide band gap. It is a promising candidate for use in silicon-based optoelectronics since its lattice resembles with silicon. It participates in various reactions with Grignard reagents.
Catalog No. 50-289-9250 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2129462KG
Cu is present in 1 (cuprous) valence state in CuCl. CuCl is a semiconductor having a wide band gap. It is a promising candidate for use in silicon-based optoelectronics since its lattice resembles with silicon. It participates in various reactions with Grignard reagents.