Learn More
AdipoGen CopperII tartrate hydrate
SDP
Catalog No. 502561409
Encompass_Preferred
Chemical. CAS 946843-80-7. Formula C4H4CuO6 . H2O. MW 211.7 . 18.0. Synthetic. Used as a chiral selector in electrophoresis. Substance for proteomics research.
Catalog No. 50-256-1409 Supplier AdipoGen Supplier No. CDXC0241G025
Chemical. CAS 946843-80-7. Formula C4H4CuO6 . H2O. MW 211.7 . 18.0. Synthetic. Used as a chiral selector in electrophoresis. Substance for proteomics research.