Learn More
Medchemexpress LLC Coproporphyrin I-15N4 | 531-14-6 | 99.2% | 658.68 g/mol | C36H38(15N)4O8 | 1 MG
SDP
Catalog No. 5000284300
Encompass_Preferred
Coproporphyrin I-15N4 is a 15N-labeled isotopically enriched standard of coproporphyrin I supplied as a solid for laboratory research. It is used as an analytical or reference standard in studies of liver function, porphyria, and metabolic profiling. The material has high isotopic enrichment and purity and should be stored protected from light at -20°C; in solution, store at -80°C for up to 6 months.
  • 15N-labeled coproporphyrin I isotopic standard.
  • High purity suitable for analytical reference (99.19%).
  • Supplied as a 1 mg solid for trace analyses.
  • Intended for research use only.
  • Store protected from light at -20°C; in solvent, -80°C up to 6 months.
Catalog No. 50-002-84300 Supplier Medchemexpress LLC Supplier No. HYW6539821MG
Coproporphyrin I-15N4 is a 15N-labeled isotopically enriched standard of coproporphyrin I supplied as a solid for laboratory research. It is used as an analytical or reference standard in studies of liver function, porphyria, and metabolic profiling. The material has high isotopic enrichment and purity and should be stored protected from light at -20°C; in solution, store at -80°C for up to 6 months.
  • 15N-labeled coproporphyrin I isotopic standard.
  • High purity suitable for analytical reference (99.19%).
  • Supplied as a 1 mg solid for trace analyses.
  • Intended for research use only.
  • Store protected from light at -20°C; in solvent, -80°C up to 6 months.