Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coproporphyrin I dihydrochloride | 69477-27-6 | 98.0% | 727.63 g/mol | C36H40Cl2N4O8 | 25 MG
SDP

Catalog No. 5000209726
Encompass_Preferred

Coproporphyrin I dihydrochloride is the dihydrochloride salt of coproporphyrin I, supplied as a solid reagent for biochemical assays and analytical reference standards. It has a reported purity of 98.0% and a molecular weight of 727.63 g/mol.

  • Solid reagent suitable for biochemical assays and analytical reference standards.
  • Reported purity 98.0%.
  • CAS number 69477-27-6 and molecular formula C36H40Cl2N4O8.
  • Available in a 25 MG pack size for laboratory use.

Catalog No. 50-002-09726 Supplier Medchemexpress LLC Supplier No. 25MGHYW073080
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coproporphyrin I dihydrochloride is the dihydrochloride salt of coproporphyrin I, supplied as a solid reagent for biochemical assays and analytical reference standards. It has a reported purity of 98.0% and a molecular weight of 727.63 g/mol.

  • Solid reagent suitable for biochemical assays and analytical reference standards.
  • Reported purity 98.0%.
  • CAS number 69477-27-6 and molecular formula C36H40Cl2N4O8.
  • Available in a 25 MG pack size for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.