Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coproporphyrin I dihydrochloride | 69477-27-6 | MFCD00013469 | 98.0% | 727.63 | C36H40Cl2N4O8 | 10 MG
SDP

Catalog No. 5000210235
Encompass_Preferred

Coproporphyrin I dihydrochloride is a biochemical reagent and endogenous porphyrin metabolite supplied as a solid for life-science research applications, including studies of liver disease and porphyria. It is commonly used as a reference material or reagent in biochemical assays.

  • High purity (98.0%) solid, purple to purplish red.
  • Molecular weight 727.63 g·mol⁻¹ and formula C36H40Cl2N4O8.
  • Soluble in DMSO (125 mg/mL); ultrasonic assistance recommended.
  • Storage: in solvent, -80°C (2 years) or -20°C (1 year); store solid dry and cool.
  • Typical applications: biochemical assays and metabolite research.

Catalog No. 50-002-10235 Supplier Medchemexpress LLC Supplier No. HYW07308010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coproporphyrin I dihydrochloride is a biochemical reagent and endogenous porphyrin metabolite supplied as a solid for life-science research applications, including studies of liver disease and porphyria. It is commonly used as a reference material or reagent in biochemical assays.

  • High purity (98.0%) solid, purple to purplish red.
  • Molecular weight 727.63 g·mol⁻¹ and formula C36H40Cl2N4O8.
  • Soluble in DMSO (125 mg/mL); ultrasonic assistance recommended.
  • Storage: in solvent, -80°C (2 years) or -20°C (1 year); store solid dry and cool.
  • Typical applications: biochemical assays and metabolite research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.