Learn More
Medchemexpress LLC Coproporphyrin III dihydrochloride | 68938-73-8 | MFCD00058927 | 99.6% | 727.63 | C36H40Cl2N4O8 | 1 MG
SDP
Catalog No. 5000281027
Encompass_Preferred
Coproporphyrin III dihydrochloride is a porphyrin derivative provided as a high-purity research reagent for biochemical and analytical applications. It is supplied in both solid and solution formats to support assays, reference standards, and method development.
  • High purity: 99.6% suitable for analytical and research use.
  • Available in solid and solution formats for experimental flexibility.
  • Multiple pack sizes: includes 1 mg, 5 mg, and 10 mg options; larger quantities by request.
  • Well-characterized: CAS 68938-73-8, molecular weight 727.63, and defined chemical formula.
  • Applicable for biochemical assays, analytical characterization, and reagent preparation.
Catalog No. 50-002-81027 Supplier Medchemexpress LLC Supplier No. HYW4121751MG
Coproporphyrin III dihydrochloride is a porphyrin derivative provided as a high-purity research reagent for biochemical and analytical applications. It is supplied in both solid and solution formats to support assays, reference standards, and method development.
  • High purity: 99.6% suitable for analytical and research use.
  • Available in solid and solution formats for experimental flexibility.
  • Multiple pack sizes: includes 1 mg, 5 mg, and 10 mg options; larger quantities by request.
  • Well-characterized: CAS 68938-73-8, molecular weight 727.63, and defined chemical formula.
  • Applicable for biochemical assays, analytical characterization, and reagent preparation.