Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Coproporphyrin III dihydrochloride | 68938-73-8 | MFCD00058927 | 99.6% | 727.63 | C36H40Cl2N4O8 | 5 MG
SDP

Catalog No. 5000281063
Encompass_Preferred

Coproporphyrin III dihydrochloride is a research-grade porphyrin derivative supplied as a purple to purplish red solid powder for in vitro microbiology and biochemical studies, including investigations of bacterial aggregation and biofilm formation.

  • High purity (99.57%) suitable for research applications.
  • Purple to purplish red solid physical form.
  • Available in small quantities including 1 mg, 5 mg, and 10 mg vials.
  • Storage: sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • CAS number 68938-73-8 for unambiguous identification.
  • For research use only; not for sale to patients.

Catalog No. 50-002-81063 Supplier Medchemexpress LLC Supplier No. HYW4121755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Coproporphyrin III dihydrochloride is a research-grade porphyrin derivative supplied as a purple to purplish red solid powder for in vitro microbiology and biochemical studies, including investigations of bacterial aggregation and biofilm formation.

  • High purity (99.57%) suitable for research applications.
  • Purple to purplish red solid physical form.
  • Available in small quantities including 1 mg, 5 mg, and 10 mg vials.
  • Storage: sealed at 4°C; in solvent: -80°C (6 months) or -20°C (1 month).
  • CAS number 68938-73-8 for unambiguous identification.
  • For research use only; not for sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.