Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Coprostan-3-Ol

Catalog No. C757810MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mg
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C757810MG 50 mg
C757850MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C757810MG Supplier Sigma Aldrich Supplier No. C757810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Coprostan-3-ol is a cholesterol derivative that can be prepared from 5+a-cholestan-3-one via reduction.It has been used as an internal standard for the determination of total amounts of cholesterol and 7-dehydrocholesterol reductase by GC/MS.

Specifications

CAS 360-68-9
Molecular Formula C27H48O
Linear Formula C27H48O
MDL Number MFCD00068259
Quantity 50 mg
Synonym 5beta-Cholestan-3beta-ol
Molecular Weight (g/mol) 388.67 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.