Learn More
Cayman Chemical Coprostnol 100mg
SDP
Catalog No. 502932940
Encompass_Preferred
A cholesterol derivative; formed via conversion of cholesterol to cholestenone then coprostanone intermediates or by direct reduction of the 5,6 double bond; commonly used as a marker of sewage contamination in soil and watersheds
Catalog No. 50-293-2940 Supplier Cayman Chemical Supplier No. 26764100 MG
A cholesterol derivative; formed via conversion of cholesterol to cholestenone then coprostanone intermediates or by direct reduction of the 5,6 double bond; commonly used as a marker of sewage contamination in soil and watersheds