Learn More
Cayman Chemical Coprostanol, Purity: ≥95%, CAS: 360-68-9, 50 mg
SDP
Catalog No. 502932941
Encompass_Preferred
A cholesterol derivative; formed via conversion of cholesterol to cholestenone then coprostanone intermediates or by direct reduction of the 5,6 double bond; commonly used as a marker of sewage contamination in soil and watersheds
Catalog No. 50-293-2941 Supplier Cayman Chemical Supplier No. 2676450 MG
A cholesterol derivative; formed via conversion of cholesterol to cholestenone then coprostanone intermediates or by direct reduction of the 5,6 double bond; commonly used as a marker of sewage contamination in soil and watersheds