Learn More
Aviva Systems Biology Corp COPS2 Antibody - N-terminal region (ARP34189_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34189_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against COPS2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human COPS2. Alternate names: ALIEN, CSN2, SGN2, TRIP15ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55292