Learn More
Aviva Systems Biology Corp COPS4 Antibody - middle region (ARP56841_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56841_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COPS4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human COPS4. Alternate names: MGC10899, MGC15160, CSN4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62006