Learn More
Aviva Systems Biology Corp COPS5 Antibody - N-terminal region (ARP32834_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32834_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COPS5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human COPS5. Alternate names: CSN5, JAB1, SGN5, MOV-34ApplicationIHC, WB. Species reactivity: Human
Catalog No. 50-014-54783