Learn More
Aviva Systems Biology Corp COPS6 Antibody - middle region (ARP59350_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59350_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COPS6. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human COPS6. Alternate names: CSN6, MOV34-34KDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-671