Learn More
Aviva Systems Biology Corp COPS8 Antibody - middle region (ARP52242_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52242_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against COPS8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human COPS8. Alternate names: COP9, CSN8, MGC1297, MGC43256, SGN8ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-669