Learn More
Medchemexpress LLC HY-N0736 5mg Medchemexpress, Coptisine (chloride) CAS:6020-18-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07365MG
Encompass_Preferred
Medchemexpress, HY-N0736 5mg Coptisine (chloride) CAS:6020-18-4 Coptisine chloride is an alkaloid from Chinese goldthread, and acts as an efficient uncompetitive IDO inhibitor with a K i value of 5.8 μM and an IC 50 value of 6.3 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3694