Learn More
Copy Cast Casting Kit
SDPSCI_ED
Catalog No. S69042A Shop All Lynn Peavey Company Products
Change view
Click to view available options
Product Type:
Copy Cast™ Casting Kit
Catalog No. Product Type
S69042A Copy Cast™ Casting Kit
1 options
This item is not returnable. View return policy
Catalog No. S69042A Supplier Lynn Peavey Company Supplier No. 09707
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Five one-gallon (3.8L) plastic reseal bags (2.5 lb. [1.13kg] of casting material), measuring cup, small bottle of hardening solution, complete instructions
Mix-and-pour convenience.
  • Made of a lab model stone-type III casting material
  • Does not require any reinforcement material
  • In most cases, a casting form is not necessary
Specifications
For Use With (Application) Ideal for impressions of footprints or tire tracks
Includes Five 1 gal. plastic resealable bags containing 2 lb. of casting material, one measuring cup
Material Lab model stone-type III casting material
Product Type Copy Cast™ Casting Kit