Learn More
LGC GENOMICS LLC (Lucigen) CopyControl Induction Solution, 25 mL
Catalog No. NC9063127
Encompass
Prepare unbiased fosmid libraries of approx. 40 kb that can be maintained at single-copy number and induced to high-copy number as needed when grown in the TransforMax EC300 E. coli.
This item is not returnable. View return policy
Catalog No. NC9063127 Supplier LGC GENOMICS LLC Supplier No. CCIS125
Prepare unbiased fosmid libraries of approx. 40 kb that can be maintained at single-copy number and induced to high-copy number as needed when grown in the TransforMax EC300 E. coli.